Learn More
Biomatik Corporation Leukocyte Immunoglobulin Like Receptor Subfamily B, Member 4 (Lilrb4) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26009
Encompass_Preferred
Leukocyte Immunoglobulin Like Receptor Subfamily B, Member 4 (Lilrb4) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Leukocyte Immunoglobulin Like Receptor Subfamily B, Member 4 (LILRB4). The Leukocyte Immunoglobulin Like Receptor Subfamily B, Member 4 (Lilrb4) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gly24~Val206. Applications: WB, IHC, ICC, IP. Target Synonyms: Leukocyte Immunoglobulin Like Receptor Subfamily B, Member 4 (LILRB4). Accession Number: Q64281. Gene ID: 14728. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9431