Learn More
Biomatik Corporation Casein Kappa (Csn3) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU29057
Encompass_Preferred
Casein Kappa (Csn3) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Casein Kappa (CSN3). The Casein Kappa (Csn3) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Gln22~Val190. Applications: WB, IHC, ICC, IP. Target Synonyms: Casein Kappa (CSN3). Accession Number: P02668. Gene ID: 281728. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9563