Learn More
Biomatik Corporation Agrin (Agrn) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26179
Encompass_Preferred
Agrin (Agrn) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Agrin (AGRN). The Agrin (Agrn) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ala1206~Thr1475. Applications: WB, IHC, ICC, IP. Target Synonyms: Agrin (AGRN). Accession Number: P25304. Gene ID: 25592. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9443