Learn More
Biomatik Corporation Gastrin (Gt) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26335
Encompass_Preferred
Gastrin (Gt) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Gastrin (GT). The Gastrin (Gt) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Asn101. Applications: WB, IHC, ICC, IP. Target Synonyms: Gastrin (GT). Accession Number: P01350. Gene ID: 2520. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9453