Learn More
Biomatik Corporation Albumin (Alb) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26643
Encompass_Preferred
Albumin (Alb) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Albumin (ALB). The Albumin (Alb) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Applications: WB, IHC, ICC, IP. Target Synonyms: Albumin (ALB). Accession Number: P07724. Gene ID: 11657. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9467