Learn More
Biomatik Corporation Cluster Of Differentiation 161 (Cd161) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26718
Encompass_Preferred
Cluster Of Differentiation 161 (Cd161) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Cluster Of Differentiation 161 (CD161). The Cluster Of Differentiation 161 (Cd161) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Lys73~Ser188. Applications: WB, IHC, ICC, IP. Target Synonyms: Cluster Of Differentiation 161 (CD161). Accession Number: Q12918. Gene ID: 3820. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9470