Learn More
Biomatik Corporation Retinol Binding Protein 4 (Rbp4) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26820
Encompass_Preferred
Retinol Binding Protein 4 (Rbp4) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Retinol Binding Protein 4 (RBP4). The Retinol Binding Protein 4 (Rbp4) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ala18~Leu201. Applications: WB, IHC, ICC, IP. Target Synonyms: Retinol Binding Protein 4 (RBP4). Accession Number: P02753. Gene ID: 5950. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9473