Learn More
Biomatik Corporation Interleukin 6 (Il6) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29173
Encompass_Preferred
Interleukin 6 (Il6) Monoclonal Antibody is a Rhesus monkey (Simian) Monoclonal Antibody to Interleukin 6 (IL6). The Interleukin 6 (Il6) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Rhesus monkey (Simian). Immunogen: Pro27~Met212. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 6 (IL6). Accession Number: P51494. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9573