Learn More
Biomatik Corporation Gonadotropin Releasing Hormone (Gnrh) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26976
Encompass_Preferred
Gonadotropin Releasing Hormone (Gnrh) Polyclonal Antibody is a Chicken (Gallus) Polyclonal Antibody to Gonadotropin Releasing Hormone (GnRH). The Gonadotropin Releasing Hormone (Gnrh) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Chicken (Gallus). Immunogen: Gln24~Ile92. Applications: WB, IHC, ICC, IP. Target Synonyms: Gonadotropin Releasing Hormone (GnRH). Accession Number: P37042. Gene ID: 770134. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9491