Learn More
Biomatik Corporation Neuromedin S (Nms) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27015
Encompass_Preferred
Neuromedin S (Nms) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Neuromedin S (NMS). The Neuromedin S (Nms) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Leu31~Gln153. Applications: WB, IHC, ICC, IP. Target Synonyms: Neuromedin S (NMS). Accession Number: Q5H8A1. Gene ID: 433292. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9493