Learn More
Biomatik Corporation Cyclin Dependent Kinase Inhibitor 2A (Cdkn2A) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27125
Encompass_Preferred
Cyclin Dependent Kinase Inhibitor 2A (Cdkn2A) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Cyclin Dependent Kinase Inhibitor 2A (CDKN2A). The Cyclin Dependent Kinase Inhibitor 2A (Cdkn2A) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC/IF, FCM. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser8~Asp156. Applications: WB, IHC, ICC/IF, FCM. Target Synonyms: Cyclin Dependent Kinase Inhibitor 2A (CDKN2A). Accession Number: P42771. Gene ID: 1029. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9495