Learn More
Biomatik Corporation Kidney Injury Molecule 1 (Kim1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27145
Encompass_Preferred
Kidney Injury Molecule 1 (Kim1) Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to Kidney Injury Molecule 1 (Kim1). The Kidney Injury Molecule 1 (Kim1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Val5~Val107. Applications: WB, IHC, ICC, IP. Target Synonyms: Kidney Injury Molecule 1 (Kim1). Accession Number: C8CLK5. Gene ID: 100303721. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9497