Learn More
Biomatik Corporation Myosin Heavy Chain 2 (Myh2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27224
Encompass_Preferred
Myosin Heavy Chain 2 (Myh2) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Myosin Heavy Chain 2 (MYH2). The Myosin Heavy Chain 2 (Myh2) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Leu1238~Ala1472. Applications: WB, IHC. Target Synonyms: Myosin Heavy Chain 2 (MYH2). Accession Number: I6L963. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9499