Learn More
Biomatik Corporation Laminin Alpha 1 (Lama1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27622
Encompass_Preferred
Laminin Alpha 1 (Lama1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Laminin Alpha 1 (LAMA1). The Laminin Alpha 1 (Lama1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ala886~Val1039. Applications: WB, IHC, ICC, IP. Target Synonyms: Laminin Alpha 1 (LAMA1). Accession Number: P25391. Gene ID: 284217. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9517