Learn More
Biomatik Corporation Transforming Growth Factor Beta 1 (Tgfb1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28534
Encompass_Preferred
Transforming Growth Factor Beta 1 (Tgfb1) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Transforming Growth Factor Beta 1 (TGFb1). The Transforming Growth Factor Beta 1 (Tgfb1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ala279~Ser390. Applications: WB, IHC, ICC, IP. Target Synonyms: Transforming Growth Factor Beta 1 (TGFb1). Accession Number: P17246. Gene ID: 59086. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9549