Learn More
Biomatik Corporation Reverse Triiodothyronine (Rt3) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29401
Encompass_Preferred
Reverse Triiodothyronine (Rt3) Monoclonal Antibody is a Universal Monoclonal Antibody to Reverse Triiodothyronine (rT3). The Reverse Triiodothyronine (Rt3) Monoclonal Antibody has been validated for the following applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Host: Mouse. Species: Universal. Applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Target Synonyms: Reverse Triiodothyronine (rT3). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9589