Learn More
Biomatik Corporation Synaptotagmin Like Protein 2 (Sytl2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU22069
Encompass_Preferred
Synaptotagmin Like Protein 2 (Sytl2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Synaptotagmin Like Protein 2 (SYTL2). The Synaptotagmin Like Protein 2 (Sytl2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Lys329~Leu880. Applications: WB, IHC, ICC, IP. Target Synonyms: Synaptotagmin Like Protein 2 (SYTL2). Accession Number: Q9HCH5. Gene ID: 54843. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9238