Learn More
Biomatik Corporation Latent Transforming Growth Factor Beta Binding Protein 1 (Ltbp1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24634
Encompass_Preferred
Latent Transforming Growth Factor Beta Binding Protein 1 (Ltbp1) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Latent Transforming Growth Factor Beta Binding Protein 1 (LTBP1). The Latent Transforming Growth Factor Beta Binding Protein 1 (Ltbp1) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Asp1415~Glu1712. Applications: WB, IHC. Target Synonyms: Latent Transforming Growth Factor Beta Binding Protein 1 (LTBP1). Accession Number: Q8CG19. Gene ID: 268977. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9358