Learn More
Biomatik Corporation Hepcidin (Hepc) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24930
Encompass_Preferred
Hepcidin (Hepc) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Hepcidin (Hepc). The Hepcidin (Hepc) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Thr24~Thr83. Applications: WB, IHC, ICC, IP. Target Synonyms: Hepcidin (Hepc). Accession Number: Q9EQ21. Gene ID: 84506. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9370