Learn More
Biomatik Corporation Caspase 1 (Casp1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25662
Encompass_Preferred
Caspase 1 (Casp1) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Caspase 1 (CASP1). The Caspase 1 (Casp1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Asn119~Asp296. Applications: WB, IHC, ICC, IP. Target Synonyms: Caspase 1 (CASP1). Accession Number: P29452. Gene ID: 12362. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9406