Learn More
Biomatik Corporation Lipolysis Stimulated Lipoprotein Receptor (Lsr) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29883
Encompass_Preferred
Lipolysis Stimulated Lipoprotein Receptor (Lsr) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Lipolysis Stimulated Lipoprotein Receptor (LSR). The Lipolysis Stimulated Lipoprotein Receptor (Lsr) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Lys335~Arg589. Applications: WB, IHC. Target Synonyms: Lipolysis Stimulated Lipoprotein Receptor (LSR). Accession Number: Q86X29. Gene ID: 51599. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9621