Learn More
Biomatik Corporation Mannose Receptor C Type 1 (Mrc1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25747
Encompass_Preferred
Mannose Receptor C Type 1 (Mrc1) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Mannose Receptor C Type 1 (MRC1). The Mannose Receptor C Type 1 (Mrc1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Leu225~Ser492. Applications: WB, IHC, ICC, IP. Target Synonyms: Mannose Receptor C Type 1 (MRC1). Accession Number: Q61830. Gene ID: 17533. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9415