Learn More
Biomatik Corporation Pulmonary Activation Regulated Chemokine (Parc) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25775
Encompass_Preferred
Pulmonary Activation Regulated Chemokine (Parc) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Pulmonary Activation Regulated Chemokine (PARC). The Pulmonary Activation Regulated Chemokine (Parc) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ala21~Ala89. Applications: WB, IHC. Target Synonyms: Pulmonary Activation Regulated Chemokine (PARC). Accession Number: P55774. Gene ID: 6362. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9417