Learn More
Biomatik Corporation Lipopolysaccharide Binding Protein (Lbp) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25992
Encompass_Preferred
Lipopolysaccharide Binding Protein (Lbp) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Lipopolysaccharide Binding Protein (LBP). The Lipopolysaccharide Binding Protein (Lbp) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gly25~Pro259. Applications: WB, IHC, ICC, IP. Target Synonyms: Lipopolysaccharide Binding Protein (LBP). Accession Number: Q61805. Gene ID: 16803. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9430