Learn More
Biomatik Corporation Involucrin (Inv) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26129
Encompass_Preferred
Involucrin (Inv) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Involucrin (INV). The Involucrin (Inv) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gln36~Leu152. Applications: WB, IHC, ICC, IP. Target Synonyms: Involucrin (INV). Accession Number: P07476. Gene ID: 3713. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9440