Learn More
Biomatik Corporation Complement C4-B (C4B) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26177
Encompass_Preferred
Complement C4-B (C4B) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Complement C4-B (C4B). The Complement C4-B (C4B) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gln126~Ser366. Applications: WB, IHC, ICC, IP. Target Synonyms: Complement C4-B (C4B). Accession Number: P01029. Gene ID: 12268. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9442