Learn More
Biomatik Corporation Neurotensin (Nt) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26370
Encompass_Preferred
Neurotensin (Nt) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Neurotensin (NT). The Neurotensin (Nt) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: His42~Ser167. Applications: WB, IHC, ICC, IP. Target Synonyms: Neurotensin (NT). Accession Number: P30990. Gene ID: 4922. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9456