Learn More
Biomatik Corporation Myosin Light Chain Kinase (Mylk) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26498
Encompass_Preferred
Myosin Light Chain Kinase (Mylk) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Myosin Light Chain Kinase (MYLK). The Myosin Light Chain Kinase (Mylk) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Phe1661~Asp1877. Applications: WB, IHC, ICC, IP. Target Synonyms: Myosin Light Chain Kinase (MYLK). Accession Number: Q15746. Gene ID: 4638. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9464