Learn More
Biomatik Corporation Alpha-Lactalbumin (Ala) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26668
Encompass_Preferred
Alpha-Lactalbumin (Ala) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Alpha-Lactalbumin (aLA). The Alpha-Lactalbumin (Ala) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Thr24~Glu141. Applications: WB. Target Synonyms: Alpha-Lactalbumin (aLA). Accession Number: P29752. Gene ID: 16770. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9468