Learn More
Biomatik Corporation Tumor Protein P53 (P53) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30692
Encompass_Preferred
Tumor Protein P53 (P53) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Tumor Protein p53 (P53). The Tumor Protein P53 (P53) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Met133~Asp393. Applications: WB. Target Synonyms: Tumor Protein p53 (P53). Accession Number: P04637. Gene ID: 7157. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9649