Learn More
Biomatik Corporation Complement Component 3 (C3) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26930
Encompass_Preferred
Complement Component 3 (C3) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Complement Component 3 (C3). The Complement Component 3 (C3) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Leu1000~Glu1326. Applications: WB, IHC, ICC, IP. Target Synonyms: Complement Component 3 (C3). Accession Number: P01024. Gene ID: 718. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9486