Learn More
Biomatik Corporation Haptoglobin (Hpt) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27052
Encompass_Preferred
Haptoglobin (Hpt) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Haptoglobin (Hpt). The Haptoglobin (Hpt) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ile104~Lys346. Applications: WB, IHC, ICC, IP. Target Synonyms: Haptoglobin (Hpt). Accession Number: P06866. Gene ID: 24464. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9494