Learn More
Biomatik Corporation Fibrillin 1 (Fbn1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27571
Encompass_Preferred
Fibrillin 1 (Fbn1) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Fibrillin 1 (FBN1). The Fibrillin 1 (Fbn1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Cys168~Arg364. Applications: WB, IHC, ICC, IP. Target Synonyms: Fibrillin 1 (FBN1). Accession Number: G3V9M6; D3ZQM5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9516