Learn More
Biomatik Corporation Myoglobin (Myo) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27830
Encompass_Preferred
Myoglobin (Myo) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Myoglobin (MYO). The Myoglobin (Myo) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Met1~Gly154. Applications: WB, IHC, ICC, IP. Target Synonyms: Myoglobin (MYO). Accession Number: P02192. Gene ID: 280695. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9525