Learn More
Biomatik Corporation Protein Kinase C Epsilon (Pkce) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27898
Encompass_Preferred
Protein Kinase C Epsilon (Pkce) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Protein Kinase C Epsilon (PKCe). The Protein Kinase C Epsilon (Pkce) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Leu403~Asp672. Applications: WB, IHC, ICC, IP. Target Synonyms: Protein Kinase C Epsilon (PKCe). Accession Number: Q02156. Gene ID: 5581. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9529