Learn More
Biomatik Corporation Integrin Alpha D (Itgad) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28220
Encompass_Preferred
Integrin Alpha D (Itgad) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Integrin Alpha D (ITGaD). The Integrin Alpha D (Itgad) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Phe886~Glu1106. Applications: WB, IHC, ICC, IP. Target Synonyms: Integrin Alpha D (ITGaD). Accession Number: Q3V0T4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9539