Learn More
Biomatik Corporation N-Acetylgalactosamine-6-Sulfatase (Galns) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28251
Encompass_Preferred
N-Acetylgalactosamine-6-Sulfatase (Galns) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to N-Acetylgalactosamine-6-Sulfatase (GALNS). The N-Acetylgalactosamine-6-Sulfatase (Galns) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Asp172~Asp381. Applications: WB, IHC, ICC, IP. Target Synonyms: N-Acetylgalactosamine-6-Sulfatase (GALNS). Accession Number: Q32KJ6. Gene ID: 292073. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9541