Learn More
Biomatik Corporation Ki-67 Protein (Ki-67) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU31531
Encompass_Preferred
Ki-67 Protein (Ki-67) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Ki-67 Protein (Ki-67). The Ki-67 Protein (Ki-67) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Thr2969~Val3119. Applications: WB, IHC, ICC, IP. Target Synonyms: Ki-67 Protein (Ki-67). Accession Number: D4A0Y6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9660