Learn More
Biomatik Corporation Immunoglobulin G4 (Igg4) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28279
Encompass_Preferred
Immunoglobulin G4 (Igg4) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Immunoglobulin G4 (IgG4). The Immunoglobulin G4 (Igg4) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Applications: WB, IHC. Target Synonyms: Immunoglobulin G4 (IgG4). Accession Number: P01861. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9543