Learn More
Biomatik Corporation Glial Fibrillary Acidic Protein (Gfap) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28753
Encompass_Preferred
Glial Fibrillary Acidic Protein (Gfap) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Glial Fibrillary Acidic Protein (GFAP). The Glial Fibrillary Acidic Protein (Gfap) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Val113~Glu211. Applications: WB, IHC, ICC, IP. Target Synonyms: Glial Fibrillary Acidic Protein (GFAP). Accession Number: P47819. Gene ID: 24387. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9551