Learn More
Biomatik Corporation Lysozyme (Lzm) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26397
Encompass_Preferred
Lysozyme (Lzm) Polyclonal Antibody is a Bovine (Bos taurus) Polyclonal Antibody to Lysozyme (LZM). The Lysozyme (Lzm) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Bovine (Bos taurus). Immunogen: Gly18~Leu147. Applications: WB, IHC. Target Synonyms: Lysozyme (LZM). Accession Number: P04421. Gene ID: 280849. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9458