Learn More
Biomatik Corporation Metallothionein 1 (Mt1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26487
Encompass_Preferred
Metallothionein 1 (Mt1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Metallothionein 1 (MT1). The Metallothionein 1 (Mt1) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Ala61. Applications: WB, IHC. Target Synonyms: Metallothionein 1 (MT1). Accession Number: P04731. Gene ID: 4489. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9462