Learn More
Biomatik Corporation Rev1 Homolog (Rev1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21339
Encompass_Preferred
Rev1 Homolog (Rev1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to REV1 Homolog (REV1). The Rev1 Homolog (Rev1) Polyclonal Antibody has been validated for the following applications: WB, ICC/IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser301~Lys478. Applications: WB, ICC/IF. Target Synonyms: REV1 Homolog (REV1). Accession Number: Q9UBZ9. Gene ID: 51455. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9202