Learn More
Biomatik Corporation Protein Tyrosine Phosphatase Receptor Type M (Ptprm) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26835
Encompass_Preferred
Protein Tyrosine Phosphatase Receptor Type M (Ptprm) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Protein Tyrosine Phosphatase Receptor Type M (PTPRM). The Protein Tyrosine Phosphatase Receptor Type M (Ptprm) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Thr1197~Ser1403. Applications: WB, IHC, ICC, IP. Target Synonyms: Protein Tyrosine Phosphatase Receptor Type M (PTPRM). Accession Number: P28827. Gene ID: 5797. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9474