Learn More
Biomatik Corporation Vitamin C (Vc) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26837
Encompass_Preferred
Vitamin C (Vc) Polyclonal Antibody is a Universal Polyclonal Antibody to Vitamin C (VC). The Vitamin C (Vc) Polyclonal Antibody has been validated for the following applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Host: Rabbit. Species: Universal. Applications: ELISA, CLIA / IHC-Fr, ICC, IP (predicted). Target Synonyms: Vitamin C (VC). Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9476