Learn More
Biomatik Corporation Osteopontin (Opn) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26851
Encompass_Preferred
Osteopontin (Opn) Polyclonal Antibody is a Rhesus monkey (Simian) Polyclonal Antibody to Osteopontin (OPN). The Osteopontin (Opn) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rhesus monkey (Simian). Immunogen: Pro18~Asn314. Applications: WB, IHC, ICC, IP. Target Synonyms: Osteopontin (OPN). Accession Number: F6V2X3. Gene ID: 704930. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9478