Learn More
Biomatik Corporation Endothelial Nos (Enos) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26913
Encompass_Preferred
Endothelial Nos (Enos) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Endothelial NOS (eNOS). The Endothelial Nos (Enos) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Arg756~Gln888. Applications: WB, IHC, ICC, IP. Target Synonyms: Endothelial NOS (eNOS). Accession Number: P29474. Gene ID: 4846. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9483