Learn More
Biomatik Corporation Caspase 8 (Casp8) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26955
Encompass_Preferred
Caspase 8 (Casp8) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Caspase 8 (CASP8). The Caspase 8 (Casp8) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC/IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser217~Asp384. Applications: WB, IHC, ICC/IF. Target Synonyms: Caspase 8 (CASP8). Accession Number: Q14790. Gene ID: 841. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9487