Learn More
Biomatik Corporation Glycogen Phosphorylase, Muscle (Pygm) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26963
Encompass_Preferred
Glycogen Phosphorylase, Muscle (Pygm) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Glycogen Phosphorylase, Muscle (PYGM). The Glycogen Phosphorylase, Muscle (Pygm) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC/IF. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Arg11~Gly187. Applications: WB, IHC, ICC/IF. Target Synonyms: Glycogen Phosphorylase, Muscle (PYGM). Accession Number: Q9WUB3. Gene ID: 19309. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9489