Learn More
Biomatik Corporation Cluster Of Differentiation 14 (Cd14) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27372
Encompass_Preferred
Cluster Of Differentiation 14 (Cd14) Polyclonal Antibody is a Pig (Sus scrofa; Porcine) Polyclonal Antibody to Cluster Of Differentiation 14 (CD14). The Cluster Of Differentiation 14 (Cd14) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Pig (Sus scrofa; Porcine). Immunogen: Thr21~Ala373. Applications: WB, IHC, ICC, IP. Target Synonyms: Cluster Of Differentiation 14 (CD14). Accession Number: A2SW51. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9506